Learn More
CD123 Mouse anti-Human, PerCP-Cy5.5, Clone: 7G3, BD
Mouse Monoclonal Antibody
142.53€ - 333.00€
Especificaciones
| Antígeno | CD123 |
|---|---|
| Clon | 7G3 |
| Aplicaciones | Flow Cytometry |
| Clasificación | Monoclonal |
| Conjugado | PerCP-Cyanine5.5 |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
15880579
|
BD Biosciences
560904 |
25 Tests |
143.00€ 142.53€ / cada uno Ahorro 0.47€ 0.33% Descuento |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
15899598
|
BD Biosciences
558714 |
100 Tests |
333.00€
cada uno |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Reacts with human CD123, the 70 kD IL-3 receptor α chain (IL-3Rα), which associates with the 120-140kDa β subunit. The β chain is shared with the receptors for interleukins IL-5 and GM-CSF. IL-3Rα is expressed on hematopoietic progenitors. This antibody has been reported to block the binding of 125I-IL-3 to high and low affinity IL-3 receptors. In functional experiments, this antibody was found to inhibit acute myeloid leukemia cell proliferation, basophil histamine release, endothelial cell-mediated IL-8 secretion, and neutrophil transmigration. This antibody has been reported to be useful for immunoprecipitation, western blot and immunofluorescent staining for flow cytometry. IL-3Rα plays an important role in hematopoietic progenitor cell growth and differentiation. At the Fifth HLDA Workshop, the human IL-3 receptor was designated CD123.
Flow Cytometry
Especificaciones
| CD123 | |
| Flow Cytometry | |
| PerCP-Cyanine5.5 | |
| RUO | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| IgG2a κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 7G3 | |
| Monoclonal | |
| Mouse | |
| Human | |
| Human IL-3Ra-transfected cells | |
| Primary | |
| IL3RA; IL-3RA; IL-3Rα; IL-3R-alpha; Interleukin-3 receptor subunit alpha |
For Research Use Only.
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.