missing translation for 'onlineSavingsMsg'
Learn More

CD127 Mouse anti-Human, FITC, Clone: HIL-7R-M21, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 560549

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 15880289

  • 409.00€ / cada uno
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

The hIL-7R-M21 monoclonal antibody specifically binds to the 60-90 kDa glycoprotein, CD127. CD127 is also known as the IL-7 receptor alpha (IL-7Rα) subunit. The IL-7 receptor complex is a heterodimer composed of CD127 and the common gamma chain (γc, CD132), shared by other cytokine receptors (IL-2R, IL-4R, IL-9R, IL-15R, and IL-21R). CD127 is expressed on thymocytes, T- and B-cell progenitors, mature T cells, and some lymphoid and myeloid cells. In vitro experiments show the expression of CD127 is down-regulated following T cell activation. Studies indicate that the IL-7 Receptor plays an important role in the proliferation and differentiation of mature T cells. Recently, it has been shown that low surface expression of CD127, in combination with intermediate to high surface expression of CD25, the α chain of the IL-2 receptor complex, can distinguish between human regulatory and conventional CD4+ T cells in human adult and cord blood, lymph nodes and thymus.

Especificaciones

Especificaciones

CD127
Monoclonal
FITC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human IL-7R Recombinant Protein
100 Tests
Immunology
Human
IgG1 κ
Flow Cytometry
HIL-7R-M21
IL-7R; IL7R; IL7RA; IL-7Rα; IL-7R-alpha; Interleukin-7 Receptor alpha
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.