missing translation for 'onlineSavingsMsg'
Learn More

CD138 Mouse anti-Human, FITC, Clone: MI15, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 561703

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 15881249

  • 210.00€ / cada uno
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

Reacts with CD138 (Syndecan-1), an 85-92kDa single chain transmembrane protein, strongly expressed on multiple-myeloma-derived cell lines and malignant plasma cell populations. It is also expressed on pre-B cells, immature B cells, and plasma cells, but not on mature circulating B-lymphocytes. Syndecan-1 is a member of the family of transmemrane heparan sulfate proteoglycans. It is also expressed on some non-hematopoietic cells, including embryonic mesenchymal cells, vascular smooth muscle cells, endothelial and neural cells. CD138 binds to many extracellular matrix proteins through its heparan sulfate side-chains, like fibronectin, collagen types I, III, and V, tenascin, thrombospondin, and antithrombin III. It is considered an extracellular matrix receptor that may serve as a co-receptor for fibroblast growth factor and related molecules. Monoclonal antibody MI15 blocks the binding of clone B-B4 but not clone DL-101 (other anti-syndecan-1 antibodies) by flow cytometric analysis. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Mouse
Clone: MI15
Isotype: IgG1 κ
Species Reactivity: Human

Flow Cytometry

Especificaciones

Especificaciones

CD138
Monoclonal
FITC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human U266 and XG-1 Myeloma Cell Lines
25 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
MI15
Syndecan; SDC; Syndecan 1; SDC1; SYND1
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.