missing translation for 'onlineSavingsMsg'
Learn More

CD16 Mouse anti-Human, FITC, Clone: 3G8, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 556618

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15888748

  • 377.00€ / cada uno
Fecha estimada de envío: 12-08-2024
para ver el stock

Descripción

Descripción

Reacts with the 50-65kDa transmembrane form of IgG Fc (FcγRIII), a human NK-cell-associated antigen. CD16 is expressed on NK cells as well as macrophages and granulocytes. Reports indicate that CD16 plays a role in signal transduction and NK cell activation. CD16 antibody blocks the binding of soluble immune complexes to granulocytes. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.

Host Species: Mouse
Clone: 3G8
Isotype: IgG1, κ
Cross Reactivity: Baboon, Cynomolgus

Flow Cytometry

Especificaciones

Especificaciones

CD16
Monoclonal
FITC
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Human polymorphonuclear leukocytes
50 Tests
Immunology
Baboon, Cynomolgus, Human, Rhesus Monkey
IgG1 κ
Flow Cytometry
3G8
FcRIII; Fc-gamma RIII; FCG3; FCGR3; FCGRIII; FcγRIII; IGFR3
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.