Learn More
CD177 Mouse anti-Human, Alexa Fluor 647, Clone: MEM-166, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 564238
Descripción
The MEM-166 monoclonal antibody specifically binds to CD177 which is also known as NB1 (Neutrophil-specific antigen), HNA-2a (Human neutrophil alloantigen 2a), and PRV1 (Polycythemia rubra vera 1). CD177 is a glycophosphatidylinositol (GPI)-anchored plasma membrane glycoprotein (56-64 kDa). CD177 is expressed on subpopulations of neutrophils, neutrophillic metamyelocytes and myelocytes from 89 to 97% of healthy individuals. It is also expressed on secondary granules. Its expression is upregulated on granulocytes stimulated with the chemotactic peptide f-met-leu-phe (fMLP). Although the function of CD177 has not been fully established, some reports suggest its possible role as receptor molecule involved in leucocyte migration.

Especificaciones
CD177 | |
Monoclonal | |
Alexa Fluor 647 | |
Mouse | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
MEM-166 | |
NB1 | |
Affinity Purified | |
RUO | |
Human | |
IgG1 κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.