missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD19 Magnetic Particles - DM Mouse anti-Human, BD IMag-DM, Clone: 4G7, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 558497
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
BD IMag™ anti-human CD19 Particles - DM are magnetic nanoparticles that have monoclonal antibody conjugated to their surfaces. These particles are optimized for the positive selection or depletion of CD19-bearing leukocytes using the BD IMag™ Cell Separation Magnet (Cat. No. 552311).
Especificaciones
CD19 | |
Monoclonal | |
BD IMag-DM | |
Human chronic lymphocytic leukemia (CLL) cells | |
RUO | |
Primary | |
The antibody-conjugated magnetic particles should be stored undiluted at 4°C. |
Flow Cytometry | |
4G7 | |
Mouse | |
5 mL | |
Immunology | |
Human | |
IgG1 κ |