missing translation for 'onlineSavingsMsg'
Learn More

CD314/NKG2D Mouse anti-Human, PE-CF594, Clone: 1D11, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 562498

351.15 EUR hasta el 2025-12-31
Use el código promocional "25339" para aplicar la oferta.


Código de producto. 15841979

  • 352.00€ / cada uno

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Edge
Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

The 1D11 monoclonal antibody specifically binds to NKG2D, a 42 kDa type II transmembrane glycoprotein that is also known as CD314 and KLRK1. NKG2D is a member of the C-type lectin family and is expressed on human NK cells. This activating receptor binds strongly to several ligands including MICA and MICB and ULBP-1, -2, and -3 proteins that are expressed by different target cell types. Different from natural cytotoxicity receptor (NCR), NKG2D expression is not confined to NK cells. It is also expressed on virtually all TCR γ/δ+ and CD8+TCR α/β+ T cells. NKG2D functions as a triggering receptor involved in natural cytotoxicity mediated by normal NK cells against a variety of tumors or normal target cells. Importantly, NKG2D can complement the role of NCR in tumor cell lysis. Remarkably, the combined maskings of NCR and NKG2D can reportedly lead to a complete inhibition of NK-mediated lysis of all tumor or normal cells. The 1D11 antibody can reportedly block or stimulate the function of NKG2D-positive cells.

This antibody is conjugated to BD Horizon™ PE-CF594, which has been developed exclusively by BD Biosciences as a better alternative to PE-Texas Red™. PE-CF594 excites and emits at similar wavelengths to PE-Texas Red™ yet exhibits improved brightness and spectral characteristics. Due to PE having maximal absorption peaks at 496nm and 564nm, PE-CF594 can be excited by the blue (488-nm), green (532-nm) and yellow-green (561-nm) lasers and can be detected with the same filter set as PE-Texas Red™ (eg 610/20-nm filter).

TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

CD314
Monoclonal
PE-CF594
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
NKL cells
50 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
1D11
CD314; KLRK1; KLR; NKG2D; NKG2-D; NK cell receptor D
Mouse
Affinity Purified
RUO
Human
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto
CD314/NKG2D Mouse anti-Human, PE-CF594, Clone: 1D11, BD > 50 Tests; PE-CF594

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado