Learn More
CD4 Mouse anti-Human, V450, Clone: L200, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 560811
Descripción
The L200 monoclonal antibody specifically binds to the human form of the 56 kDa transmembrane glycoprotein, CD4, present on the T-helper/inducer subset of normal human donor peripheral blood lymphocytes. The L200 antibody also crossreacts with a subset of CD3-positive peripheral blood lymphocytes, but not monocytes, of both Rhesus and Cynomolgus Macaque monkeys. Crossreactivity on both lymphocytes and monocytes (weak) from Baboons is also observed. The distribution on lymphocytes is similar for both human and monkey cells, with the majority of CD4-positive lymphocytes being CD8-negative and lacking reactivity with antibodies to B- or NK-cell markers.
Especificaciones
CD4 | |
Monoclonal | |
Violet Fluor 450 (V450) | |
Mouse | |
Affinity Purified | |
RUO | |
Baboon, Cynomolgus, Rhesus Monkey | |
IgG1 κ |
Flow Cytometry | |
L200 | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
Human HPB-ALL Cell Line | |
50 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |