Learn More
CD45RA Mouse anti-Human, PE, Clone: HI100, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 555489
Detalles adicionales : Peso : 0.02000kg
Description
The HI100 monoclonal antibody specifically binds to the 220 kDa isoform of the human leukocyte common antigen, CD45RA. CD45RA is expressed on approximately 40-50% of peripheral CD4+ T cells, 50% of peripheral CD8+ T cells and on a portion of B cells and monocytes. The CD45RA antigen is expressed by naοve and activated T cells. CD45RA-specific antibodies are useful for the study of the suppressor/inducer subpopulation of CD4+ lymphocytes.
Specifications
CD45RA | |
Monoclonal | |
PE | |
Mouse | |
100 Tests | |
Immunology | |
Human | |
IgG2b κ |
Flow Cytometry | |
HI100 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.