Learn More
CD49a Mouse anti-Human, PE, Clone: SR84, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 559596
Descripción
Reacts with a type I transmembrane molecule of approximately 200 kDa. CD49a is also known as very late antigen 1 α subunit (VLA-1) of the integrin family of cell adhesion molecules. CD49a associates with the integrin β 1 subunit (CD29) to form the α 1/ β 1 heterodimer (CD49a/CD29), and serves as receptor for collagen and laminin-1. It is expressed on activated T cells, monocytes, neuronal cells and smooth muscle cells. CD49a has been reported to play a role in cell attachment during the development of both the central and peripheral nervous systems.
Especificaciones
| CD49a | |
| Monoclonal | |
| PE | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| P-Selectin IgG1 Fusion Protein | |
| 100 Tests | |
| Immunology | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| SR84 | |
| Integrin α1 Chain | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.