missing translation for 'onlineSavingsMsg'
Learn More

CD49f Rat anti-Human, Alexa Fluor 647, Clone: GoH3, BD

Rat Monoclonal Antibody

Marca:  BD Biosciences 562473

Detalles adicionales : Peso : 0.00800kg

 Ver más versiones de este producto

Código de producto. 15891949

  • 740.00€ / cada uno
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

The GoH3 monoclonal antibody specifically binds to CD49f which is also known as integrin α6 chain. CD49f is a ∽150 kDa type I transmembrane glycoprotein that belongs to the integrin alpha chain family of extracellular matrix and cell adhesion receptors. The integrin α6 subunit associates with the integrin β1 chain (CD29) to form VLA-6 and with the integrin β4 chain (CD104) to form the integrin α6β4 complex, also known as the laminin and kalinin receptor. CD49f is expressed mainly on T cells, monocytes, platelets, epithelial cells, endothelial cells, perineural cells, and trophoblasts of placenta. GoH3 recognizes an extracellular epitope of integrin α6 on human, mouse and bovine cells. GoH3 has been reported to block the binding of integrin α6 to laminin P1 and E8 fragments.

Especificaciones

Especificaciones

CD49f
Monoclonal
Alexa Fluor 647
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Mouse mammary tumor cells
100 Tests
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
GoH3
ITGA6; IThe GoH3 monoclonal ntegrin alpha-6; Integrin α6 chain; VLA-6; ITA6
Rat
Affinity Purified
RUO
Human
IgG2a κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.