Learn More
CD56 Mouse anti-Human, BUV395, Clone: NCAM16.2, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 563555
Detalles adicionales : Peso : 0.01000kg
Descripción
The NCAM16.2 monoclonal antibody specifically binds to human CD56. It recognizes an extracellular immunoglobulin-like domain common to 120, 140, and 180 kDa forms of CD56, also known as the neural cell adhesion molecule (NCAM), NKH1 or MSK39. The CD56 antigen is expressed on approximately 10% to 25% of peripheral blood lymphocytes. It is present on essentially all resting and activated CD16+ natural killer (NK) lymphocytes and approximately 5% of CD3+ peripheral blood lymphocytes. CD3+ CD56+ T lymphocytes comprise a unique subset of cytotoxic T lymphocytes that mediates non-major histocompatibility complex (MHC)-restricted cytotoxicity. CD56 antigen density on NK lymphocytes increases upon cellular activation. The CD56 antigen is involved in neuronal homotypic cell adhesion and cell differentiation during embryogenesis. CD16+ CD56+ NK cells demonstrate reciprocal transfer of an activation state with dendritic cells.
The antibody was conjugated to BD Horizon™ BUV395 which has been exclusively developed by BD Biosciences as an optimal dye for use on a 355nm laser equipped instrument. With an Ex Max at 348nm and an Em Max at 395nm, this dye has virtually no spillover into any other detector. BD Horizon™ BUV395 can be excited with a 355nm laser and detected with a 379/28 filter.
Especificaciones
CD56 | |
Monoclonal | |
Brilliant Ultraviolet 395 | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Immunoaffinity-enriched adult human brain NCAM | |
25 Tests | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Flow Cytometry | |
NCAM16.2 | |
NCAM1; NCAM-1; NCAM; Leu-19; Neural cell adhesion molecule 1; NKH1; MSK39 | |
Mouse | |
Affinity Purified | |
RUO | |
Human | |
IgG2b κ |
For Research Use Only.