Learn More
CD8 Mouse anti-Human, PE-Cy5, Clone: HIT8A, BD
Mouse Monoclonal Antibody
545.00€
Especificaciones
Antígeno | CD8 |
---|---|
Clon | HIT8A |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Conjugado | PE-Cyanine5 |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15888318
|
BD Biosciences
555636 |
100 Tests |
545.00€
cada uno |
Fecha estimada de envío: 21-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Reacts with the α subunit (32kDa) of the two-chain complex. CD8 molecule binds to HLA class I molecules during interaction of CD8+ T cells with antigen-presenting cells or with target cells. CD8 is expressed on T cytotoxic/suppressor cell populations. HIT8a stains approximately 13 - 48% of peripheral blood lymphocytes and 80% of thymocytes, as well as a subset of NK cells. Clones HIT8a and RPA-T8 are not cross-blocking. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Mouse
Clone: HIT8a
Isotype: IgG1 κ
Species Reactivity: Human
Flow Cytometry
Especificaciones
CD8 | |
Flow Cytometry | |
PE-Cyanine5 | |
RUO | |
Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
HIT8A | |
Monoclonal | |
Mouse | |
Human | |
IgG1 κ | |
Affinity Purified |
For Research Use Only.