Learn More
Complement Factor H Monoclonal Antibody (OX-24), Invitrogen™
Mouse Monoclonal Antibody
Marque: Invitrogen MA170057
360.01 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alert:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Description
MA1-70057 detects Complement Factor H in human samples. MA1-70057 has been successfully used in flow cytometry, ELISA, immunoprecipitation and Western blot procedures. The MA1-70057 immunogen is human complement Factor H. Store at 4oC. For long term storage, aliquot and freeze unused portion at -20oC in volumes appropriat for single usage. Avoid freeze/thaw cycles.
Factor H is a serum protein acting as an inhibitor in the complement activation system. I activates the C3/C5 convertase by dissociating its subunits and also acts as a cofactor for the enzyme activity of Factor I.
Spécification
Complement Factor H | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
P08603 | |
CFH | |
Human complement Factor H. | |
250 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunoprecipitation, Western Blot | |
OX-24 | |
Unconjugated | |
CFH | |
adrenomedullin binding protein; age-related maculopathy susceptibility 1; AHUS1; AMBP1; ARMD4; ARMS1; beta-1H; beta-1-H-globulin; Cfh; CFHL3; complement component factor H; complement factor H; complement factor H related protein 3A4/5G4; complement factor H, isoform b; complement inhibitory factor H; factor H; factor H-like 1; FH; FHL1; FHL1 antibody; H factor 1; H factor 1 (complement); H factor 2 (complement); HF; HF1; HF2; HGNC:4883; HUS; MGC88246; Mud-1; NOM; platelet complement factor H; protein beta-1-H; RP1-177P10.1; Sas1; Sas-1 | |
Mouse | |
Protein G | |
RUO | |
3075 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Promotions
-
Multiple Manufacturers
Más descubrimientos, menos costes
Oferta ampliada hasta el 28 de marzoPromo Code: 24111Expires: 2025-03-28
New Offer Ahorre ahora
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.