missing translation for 'onlineSavingsMsg'
Learn More
Learn More
dCMP deaminase, Mouse, Clone: 4B9, Abnova™
Mouse monoclonal antibody raised against a partial recombinant DCTD.
Marca: Abnova H00001635-M01.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene catalyzes the deamination of dCMP to dUMP, the nucleotide substrate for thymidylate synthase. The encoded protein is allosterically activated by dCTP and inhibited by dTTP, and is found as a homohexamer. This protein uses zinc as a cofactor for its activity. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq
Sequence: RTAENKLDTKYPYVCHAELNAIMNKNSTDVKGCSMYVALFPCNECAKLIIQAGIKEVIFMSDKYHDSDEATAARLLFNMAGVTFRKFIPKCSKIVIDFDSINSRPSQKLQEspecificaciones
dCMP deaminase | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_001921.2 | |
DCTD | |
DCTD (NP_001912.2, 69 a.a. ∼ 178 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Endocrine/Metabolism | |
1635 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG3 κ |
ELISA, Immunohistochemistry (PFA fixed), KnockDown, Western Blot | |
4B9 | |
Mouse monoclonal antibody raised against a partial recombinant DCTD. | |
DCTD | |
MGC111062 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido