missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DEPDC7 Monoclonal Antibody (4D9), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA522853
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Peptide Sequence: NPSEFKLQKE SDNRMVVKRI FSKAIVDNKN LSKGKTDLLV LFLMDHQKDV FKIPGTLHKI VSVKLMAIQN GRDPNRDAGY IYCQRIDQRD YSNNTEKTTK DELLNLL
DEPDC7 Monoclonal antibody specifically detects DEPDC7 in Human samples. It is validated for ELISA, Immunoprecipitation.Especificaciones
DEPDC7 | |
Monoclonal | |
1 mg/mL | |
PBS with no preservative; pH 7.4 | |
Q96QD5 | |
DEPDC7 | |
DEPDC7 (NP_631899, 393 a.a. ∽ 499 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Immunoprecipitation | |
4D9 | |
Unconjugated | |
DEPDC7 | |
AV216087; DEP domain containing 7; DEP domain-containing protein 7; Depdc7; dJ85M6.4; dJ85M6.4 (novel 58.3 KDA protein); novel 58.3 KDA protein; Protein TR2/D15; RGD1309720; TR2 | |
Mouse | |
Affinity chromatography | |
RUO | |
91614 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |