missing translation for 'onlineSavingsMsg'
Learn More
Learn More
glyceraldehyde-3-phosphate dehydrogenase, Mouse, Clone: 1G5, Abnova™
Mouse monoclonal antibody raised against a partial recombinant GAPDH.
Marca: Abnova H00002597-M03.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The product of this gene catalyzes an important energy-yielding step in carbohydrate metabolism, the reversible oxidative phosphorylation of glyceraldehyde-3-phosphate in the presence of inorganic phosphate and nicotinamide adenine dinucleotide (NAD). The enzyme exists as a tetramer of identical chains. Many pseudogenes similar to this locus are present in the human genome. [provided by RefSeq
Sequence: GKLTGMAFRVPTANVSVVDLTCRLEKPAKYDDIKKVVKQASEGPLKGILGYTEHQVVSSDFNSDTHSSTFDAGAGIALNDHFVKLISWYDNEFGYSNRVVDLMAHMASKEEspecificaciones
glyceraldehyde-3-phosphate dehydrogenase | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_002046 | |
GAPDH | |
GAPDH (NP_002037, 226 a.a. ∼ 335 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Stem Cell Biology | |
2597 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG1 κ |
ELISA, Immunoprecipitation, Western Blot | |
1G5 | |
Mouse monoclonal antibody raised against a partial recombinant GAPDH. | |
GAPDH | |
G3PD/GAPD/MGC88685 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Human | |
Antibody |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido