missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GBX2 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA518561
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 8,000.
Altered expression of GBX2, part of the homeobox-containing human family of DNA-binding transcription factors, is associated with therapy failure and death in patients with multiple types of cancer.Especificaciones
GBX2 | |
Polyclonal | |
Unconjugated | |
GBX2 | |
D130058E05Rik; gastrulation and brain-specific homeobox protein 2; gastrulation brain homeobox 2; Gbx2; Gbx-2; Homeobox protein GBX-2; LOW QUALITY PROTEIN: homeobox protein GBX-2; MMoxA; Stimulated by retinoic acid gene 7 protein; Stra7 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
2637 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
P52951 | |
GBX2 | |
Synthetic peptide sequence (QGKDESKVEDDPK) corresponding to the internal amino acids of GBX2 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido