missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Granzyme B Mouse, Unlabeled, Clone: 2CF/F5, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 550558
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The primary mechanism by which cytotoxic T cells eliminate virally infected cells is by granule exocytosis. The release of cytotoxic granule contents by cytotoxic T lymphocytes (CTL) triggers apoptotic target cell death. CTL granules contain a poreforming
Host Species: Mouse
Clone: C92-605
Isotype: IgG2a
Species Reactivity: Human
Western Blotting
Especificaciones
Granzyme B | |
Monoclonal | |
0.5mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C |
Western Blot | |
2CF/F5 | |
Unconjugated | |
Mouse | |
50 μg | |
Cell Biology | |
Human, Rat | |
IgG2a |
For Research Use Only.