Learn More
anti-IgG1 κ, Mouse , Clone: MOPC-31C, BD
Marca: BD Biosciences 557273
Detalles adicionales : Peso : 0.01000kg
Descripción
The C48-6 antibody is specific for keyhole limpet hemocyanin (KLH).
Host Species: Mouse
Clone: MOPC-31C
Isotype: IgM κImmunogen: Trinitrophenol-KLH
ELISA Standard, Flow Cytometry, Immunohistochemistry (Formalin-Antigen Retrieval Required), Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Isotype Control
Especificaciones
ELISA, Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
0.5mg/mL | |
Mouse | |
Affinity Purified | |
Undiluted at 4°C |
MOPC-31C | |
Unconjugated | |
Trinitrophenol-KLH | |
0.5 mg | |
IgG1 κ |