Learn More
anti-IgG2a κ, Rat, APC, Clone: R35-95, Isotype Control, BD
453.00€ - 600.00€
Especificaciones
Clon | R35-95 |
---|---|
Aplicaciones | Flow Cytometry |
Conjugado | APC |
Especie del huésped | Rat |
Inmunógeno | Mouse Pooled Immunoglobulin |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15877478
|
BD Biosciences
553932 |
0.1 mg |
600.00€
cada uno |
Fecha estimada de envío: 12-08-2024
Inicie sesión para ver el stock |
|||||
15866548
|
BD Biosciences
551139 |
100 Tests |
453.00€
cada uno |
Fecha estimada de envío: 19-08-2024
Inicie sesión para ver el stock |
|||||
Descripción
The R35-95 hybridoma was generated by hybridization of Y3 myeloma cells with spleen cells from LOU rats immunized with mouse immunoglobulins. The R35-95 hybridoma produces rat IgG2a, κ immunoglobulin that has no measurable reactivity with mouse immunoglobulins. The R35-95 immunoglobulin was selected as an isotype control following screening for low background binding on a variety of mouse and human tissues. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only.
Host Species: Rat
Clone: R35-95
Isotype: IgG2a κ
Immunogen: Mouse Pooled Immunoglobulin
Flow Cytometry, Intracellular Staining, Isotype Control
Especificaciones
R35-95 | |
APC | |
Mouse Pooled Immunoglobulin | |
Affinity Purified |
Flow Cytometry | |
Rat | |
IgG2a κ | |
Undiluted at 4°C |