Learn More
IgM Rat anti-Mouse, HRP, Clone: II/41, BD
Rat Monoclonal Antibody
Marca: BD Biosciences 550588
Detalles adicionales : Peso : 0.01000kg
Descripción
The II/41 clone has been reported to react specifically with mouse IgM of Igh-C[a] and Igh-C[b] haplotypes. It has been reported not to react with other Ig isotypes. In addition, the II/41 clone has been reported not to stimulate B-cell proliferation.
Host Species: Rat
Clone: II/41
Isotype: IgG2a κ
Species Reactivity: Mouse
ELISA
Especificaciones
IgM | |
Monoclonal | |
HRP | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
ELISA | |
II/41 | |
Aqueous buffered solution containing BSA and ProClin™150 as preservative. | |
Not reported | |
1 mL | |
Immunology | |
Mouse | |
IgG2a κ |
For Research Use Only.