missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human Kappa Light Chain Secondary Antibody, Invitrogen™
Mouse Monoclonal Secondary Antibody
Marca: Invitrogen MA512117
216.30 EUR hasta el 2025-03-16
Use el código promocional "24070" para aplicar la oferta.
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
By Western blot, MA5-12117 detects Human Ig kappa light chain at ∽25kD in human samples. By ELISA and WB, MA5-12117 does not react with human Ig lambda light chain.
Anti-Human secondary antibodies are affinity-purified antibodies with well-characterized specificity for human immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
Especificaciones
Human Kappa Light Chain | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
Mouse | |
Protein A/G | |
RUO | |
Human | |
Liquid |
ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
L1C1 | |
Unconjugated | |
Immunoglobulin kappa light chain, Ig kappa light chain, Ig kappa, kappa light chain, Igk, Iglc | |
B lymphoma cells | |
500 μL | |
Secondary | |
4° C | |
IgG1 κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Mouse anti-Human Kappa Light Chain Secondary Antibody, Invitrogen™ > 500 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido