missing translation for 'onlineSavingsMsg'
Learn More

Laminin beta-1 Monoclonal Antibody (2D9G5), Invitrogen™

Mouse Monoclonal Antibody

Marca:  Invitrogen MA515467

Código de producto. 11335443

  • 453.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

MA5-15467 targets LAMB1 in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15467 immunogen is purified recombinant fragment of LAMB1 (aa31-270) expressed in E. Coli. <br>.

Laminins are large heterotrimeric, noncollagenous glycoproteins composed of alpha, beta, and gamma chains. They are ubiquitously present in basement membrane (BM) along with entactin/nidogen (EN), collagen type IV (CIV), and large heparan sulfate proteoglycan (HSPG), which interact specifically with each other to form a continuous and regular BM. Alterations of BM integrity, from local discontinuities up to complete loss, are described in many types of human and animal epithelial neoplasms.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

Laminin beta-1
Monoclonal
Conc. Not Determined
ascites with 0.03% sodium azide
P07942
LAMB1
Purified recombinant fragment of LAMB1 (aa31-270) expressed in E. Coli.
RUO
3912
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
ELISA, Immunohistochemistry, Western Blot
2D9G5
Unconjugated
LAMB1
C77966; C80098; C81607; CLM; D130003D08Rik; I79_017505; LAMB1; Lamb-1; LAMB1; basement membrane protein; Lamb1-1; laminin B1; laminin B1 chain; laminin B1 subunit 1; laminin beta 1; laminin beta-1 chain; laminin subunit beta 1; laminin subunit beta-1; laminin, beta 1; laminin-1 subunit beta; Laminin-10 subunit beta; laminin-12 subunit beta; Laminin-2 subunit beta; laminin-6 subunit beta; Laminin-8 subunit beta; LIS5; LOW QUALITY PROTEIN: laminin subunit beta-1; MGC142015
Mouse
100 μL
Primary
Human
Antibody
IgG2a
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales