missing translation for 'onlineSavingsMsg'
Learn More
Learn More
myotubularin 1, Mouse, Clone: 1C10, Abnova™
Mouse monoclonal antibody raised against a partial recombinant MTM1.
Marca: Abnova H00004534-M01.100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This gene encodes a dual-specificity phosphatase that acts on both phosphotyrosine and phosphoserine. It is required for muscle cell differentiation and mutations in this gene have been identified as being responsible for X-linked myotubular myopathy. [provided by RefSeq
Sequence: MASASTSKYNSHSLENESIKRTSRDGVNRDLTEAVPRLPGETLITDKEVIYICPFNGPIKGRVYITNYRLYLRSLETDSSLILDVPLGVISRIEKMGGATEspecificaciones
myotubularin 1 | |
Monoclonal | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
BC030779 | |
MTM1 | |
MTM1 (AAH30779, 1 a.a. ∼ 100 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody |
ELISA, Immunohistochemistry (PFA fixed), KnockDown, Western Blot | |
1C10 | |
Mouse monoclonal antibody raised against a partial recombinant MTM1. | |
MTM1 | |
CNM/MTMX/XLMTM | |
Mouse | |
Affinity Purified | |
RUO | |
4534 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
IgG2a κ |