Learn More
Nek2 Mouse anti-Human, Unlabeled, Clone: 20, BD
Mouse Monoclonal Antibody
Marca: BD Biosciences 610593
Detalles adicionales : Peso : 0.01000kg
Descripción
Reversible protein phosphorylation is critical for progression through the cell cycle and mitosis. In Aspergillus nidulans, the nima gene (never itosis) encodes a protein kinase that is essential for mitosis. Three human genes (nek1, 2, and 3 [Nima-related kinase]) with significant homology to the A. nidulans nima have been reported. The nek2 gene encodes a protein of 445 amino acids and, like its fungal homolog, its expression is regulated throughout the cell cycle. In Hela cells, Nek2 activity and expression are low during M and G1 phases of the cell cycle. However, both parameters increase during S phase and mitosis. In addition, Nek2 phosphorylates protein substrates exclusively at serine and threonine residues. Thus, like its fungal homolog, Nek2 may be a crucial element in controlling the cell's entry into S phase and mitosis.
This antibody is routinely tested by western blot analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.
Especificaciones
Nek2 | |
Monoclonal | |
250μg/mL | |
Nima Related Kinase 2 | |
Mouse | |
Affinity Purified | |
RUO | |
Primary | |
Store undiluted at -20°C. |
Western Blot | |
20 | |
Unconjugated | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
50 μg | |
Cell Biology | |
Human | |
IgG1 |
For Research Use Only.