missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nop5/Nop6 Mouse anti-Yeast, Clone: 37C12, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA110028
Detalles adicionales : Peso : 0.10000kg
Descripción
Nop5p, or Nop58p, is localized in the nucleolus and required for pre-18S rRNA processing in Saccharomyces cerevisiae.Especificaciones
Nop5/Nop6 | |
Monoclonal | |
Unconjugated | |
NOP58, NOP6, YOR310C, NOP58, Nucleolar protein 58, Nucleolar protein 5, Nucleolar protein 6 | |
Mouse | |
500 μL | |
Primary | |
Yeast | |
Antibody | |
IgG3 |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot | |
37C12 | |
NOP58 | |
NOP58, NOP6 | |
Yeast nuclear preparation. | |
RUO | |
851313, 854487 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |