Learn More
Nucleoporin p62 Mouse, Unlabeled, Clone: 53, BD
Mouse Monoclonal Antibody
343.00€ - 670.00€
Especificaciones
Antígeno | Nucleoporin p62 |
---|---|
Clon | 53 |
Concentración | 250μg/mL |
Aplicaciones | Western Blot |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15865489
|
BD Biosciences
610497 |
50 μg |
343.00€
1 set |
Fecha estimada de envío: 21-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15875489
|
BD Biosciences
610498 |
150 μg |
670.00€
1 set |
Fecha estimada de envío: 13-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
The nuclear pore complex (NPC) acts as a gate to mediate active transport of proteins and RNA into and out of the nucleus. Proteins actively transported into the nucleus through the NPC require specific nuclear localization sequences. Many of these nucleoporins contain N-acetylglucosamine (GlcNAc) residues that are O-linked to serine or threonine. p62 is the best characterized member of a group of nucleoporins that line the central region of the NPC. A tightly associated complex is formed by p62 and two other nucleoporins, p54 and p58. p54 binds to a carboxy-terminal coiled-coil domain of p62 and p58 binds to a dimer of p54. The amino-terminal domain of p62 contains a series of XFXFX repeats and is joined to the coiled-coil domain by a threonine-rich linker segment. The major role of p62 is maintenance of the structural integrity of NPCs.
Especificaciones
Nucleoporin p62 | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG2b | |
Affinity Purified |
53 | |
Western Blot | |
Unconjugated | |
RUO | |
Chicken, Human, Mouse, Rat | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.