missing translation for 'onlineSavingsMsg'
Learn More

Opsin 5 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA534041

Detalles adicionales : Peso : 0.01000kg

Código de producto. 15372331

  • 470.00€ / 50 microgramos
Fecha estimada de envío: 12-09-2024
para ver el stock

Descripción

Descripción

Percent identity with other species by BLAST analysis: Human, Gorilla, Monkey, Marmoset, Dog, Bovine, Elephant, Horse, Rabbit (100%) Gibbon, Mouse (94%) Bat (81%).

Opsins are members of the guanine nucleotide-binding protein (G protein)-coupled receptor superfamily. This opsin gene is expressed in the eye, brain, testes, and spinal cord. This gene belongs to the seven-exon subfamily of mammalian opsin genes that includes peropsin (RRH) and retinal G protein coupled receptor (RGR). Like these other seven-exon opsin genes, this gene may encode a protein with photoisomerase activity. Alternative splicing results in multiple transcript variants encoding different isoforms. Detected in the eye, testis, brain, retina and cell lines derived from neural retina.
Especificaciones

Especificaciones

Opsin 5
Polyclonal
Unconjugated
OPN5
DJ402H5.1; G protein-coupled receptor 136; Gpr136; G-protein coupled receptor 136; G-protein coupled receptor PGR12; GRP136; Neuropsin; OPN5; opsin 5; opsin-5; Pgr12; TMEM13; Transmembrane protein 13
Rabbit
Antigen affinity chromatography
RUO
100055886, 100353027, 100856777, 221391
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin)
1 mg/mL
PBS with 0.1% sodium azide
Q6U736
OPN5
Synthetic 16 amino acid peptide from C-terminus of human OPN5.
50 μg
Primary
Canine, Equine, Human, Non-human Primate, Rabbit
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales