missing translation for 'onlineSavingsMsg'
Learn More

PKCι Mouse, Unlabeled, Clone: 23, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 610175

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15805309

  • 433.00€ / 1 set
Fecha estimada de envío: 30-08-2024
para ver el stock

Descripción

Descripción

The Protein Kinase C (PKC) family of homologous serine/threonine protein kinases is involved in a number of processes such as growth, differentiation, and cytokine secretion. At least eleven isozymes have been described. These proteins are products of multiple genes and alternative splicing. PKC consists of a single polypeptide chain containing four conserved regions (C) and five variable regions(V). The N-terminal half containing C1, C2, V1, and V2 constitutes the regulatory domain and interacts with the PKC activators Ca2+, phospholipid, diacylglycerol, or phorbol ester. However, the novel PKC (nPKC) subfamily members (δ, ε, η and θ isoforms) and the atypical PKC (aPKC) subfamily members (ζ, ι and γ isoforms) are Ca2+ independent and lack the C2 domain. The aPKC members are unique in that their activity is independent of diacylglycerols and phorbol esters. They also lack one repeat of the cysteine-rich sequences that are conserved in cPKC and nPKC. The C-terminal region of PKC contains the catalytic domain. The PKC pathway represents a major signal transduction system that is activated following ligand-stimulation of transmembrane receptors by hormones, neurotransmitters, and growth factors. PKCι was isolated from a human kidney cDNA library. It is highly expressed in brain and lung, but it is also seen in many other tissues at lower levels. PKCι shows the most similarity to PKCζ , 72% overall identity. These two enzymes share a highly conserved pseudosubstrate sequence, the absence of a Ca2+-binding region, and the presence of only one zinc finger-like domain. It is thought that PKC? has a role in the secretory response to nutrients.

Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blotting

Especificaciones

Especificaciones

PKCι
Monoclonal
250μg/mL
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
50 μg
Cell Biology
Canine, Chicken, Human, Mouse, Rat
IgG2b
Western Blot
23
Unconjugated
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.