missing translation for 'onlineSavingsMsg'
Learn More

ROCK-I Mouse, Unlabeled, Clone: 46, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 611136

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15895809

  • 343.00€ / cada uno
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

ROCK-I is a Rho-associated serine/threonine kinase isozyme that mediates RhoA-induced assembly of focal adhesions and actin stress fibers. It contains an N-terminal kinase domain, a central 600 amino acid long coiled-coil region, a C-terminal pleckstrin homology region (PH) and a Cys-rich zinc finger motif. The ROCK-I kinase domain is approximately 90% identical to that of ROCK-II. ROCK-I binds GTP-bound Rho through a Rho-binding domain (RBD). As a result, the kinase activity of ROCK-I is moderately stimulated. The ROCK isozymes regulate cell contractility through phosphorylation of the myosin light chain. This effect results from either the inhibition of the myosin phosphatase or by direct phosphorylation of the myosin light chain, thus bypassing the myosin light chain kinase. In addition, ROCK-I activates the ubiquitously expressed Na-H exchanger (NHE1) via a number of mechanisms including RhoA. NHE1 may mediate ROCK-I-induced changes in the actin cytoskeleton. Therefore, ROCK-I plays an important role in the regulation of focal adhesion and stress fiber formation.

Host Species: Mouse
Clone: 46
Isotype: IgG1
Species Reactivity: Mouse
Immunogen: Mouse ROCK-I aa. 906-1012
Formula Weight [Chemical]: 160kDa

Immunofluorescence, Western Blotting

Especificaciones

Especificaciones

ROCK-I
Monoclonal
250μg/mL
ROKβ
Mouse
Affinity Purified
RUO
Primary
Store undiluted at -20°C.
Western Blot
46
Unconjugated
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide.
Mouse ROCK-I aa. 906-1012
50 μg
Cell Biology
Human, Mouse, Rat
IgG1
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only.