Learn More
TCP-1 beta Rat anti-Bovine, Canine, Hamster, Human, Mouse, Non-human primate, Ovine, Porcine, Rabbit, Rat, Clone: PK/8/4/4i/2f, Invitrogen™
Rat Monoclonal Antibody
Marca: Invitrogen MA190088
Detalles adicionales : Peso : 0.01000kg
Descripción
MA1-90088 detects TCP1 beta from mouse samples. MA1-90088 has been successfully used in Western blot applications. The MA1-90088 immunogen is: TCP1 beta peptide conjugated to purified tuberculin protein derivative (PPD) (Mouse).
TCP-1 beta is a molecular chaperone that is member of the chaperonin containing TCP1 complex (CCT), also known as the TCP1 ring complex (TRiC). The CCT complex consists of two identical stacked rings, each containing eight different proteins. Unfolded polypeptides enter the central cavity of the complex and are folded in an ATP-dependent manner. The complex folds various proteins, including actin and tubulin. Alternate transcriptional splice variants of the TCP-1 beta gene have been observed but not thoroughly characterized. A disease associated with TCP-1 beta protein dysfunction include epididymo-orchitis.Especificaciones
TCP-1 beta | |
Monoclonal | |
0.34 mg/mL | |
CCT2 | |
AI528772, CCT, Cct1, Ccta, TRic, Tcp-1, Tp63, c-cpn, p63 | |
Rat | |
Protein G | |
RUO | |
100157776, 100216334, 100759253, 101102222, 10576, 12461, 299809, 474445, 505313 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
PK/8/4/4i/2f | |
Unconjugated | |
Q3ZBH0, P78371, P80314, Q5XIM9 | |
CCT2 | |
TCP1 beta peptide conjugated to purified tuberculin protein derivative (PPD) (Mouse). | |
50 μg | |
Primary | |
Bovine, Canine, Hamster, Human, Mouse, Non-human Primate, Ovine, Porcine, Rabbit, Rat | |
Antibody | |
IgG2b |