missing translation for 'onlineSavingsMsg'
Learn More

TER-119/Erythroid Cells Rat anti-Mouse, Unlabeled, Clone: TER-119, BD

Rat Monoclonal Antibody

Marca:  BD Biosciences 553671

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15877328

  • 480.00€ / cada uno
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

The TER-119 antibody reacts with a 52kDa molecule associated with glycophorin A on cells of the erythroid lineage in embryonic yolk sac, fetal liver, newborn liver, adult bone marrow, adult peripheral blood, and adult lymphoid organs. The TER-119 antigen is expressed on erythroid cells from pro-erythroblast through mature erythrocyte stages, but not on cells with BFU-E or CFU-E activities. The TER-119 epitope is not detected on hematopoietic stem cells, lymphoid cells, myeloid cells, or erythroleukemia lines. The TER-119 mAb is a component of the “lineage” cocktail used in studies of hematopoietic progenitors to detect, or deplete cells committed to the hematopoietic lineages. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Host Species: Rat
Clone: TER-119
Isotype: IgG2b, κ
Species Reactivity: Mouse
Immunogen: Mouse Fetal Liver

Immunohistochemistry, Immunohistochemistry (Frozen)

Especificaciones

Especificaciones

TER-119/Erythroid Cells
Monoclonal
0.5mg/mL
Lymphocyte antigen 76; Ly76; Ly-76; TER-119; Ter119
Rat
Affinity Purified
RUO
Primary
Store undiluted at 4°C
Flow Cytometry
TER-119
Unconjugated
Aqueous buffered solution containing ≤0.09% sodium azide.
Mouse Fetal Liver
0.5 mg
Immunology
Mouse
IgG2b κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.