missing translation for 'onlineSavingsMsg'
Learn More

TNF Mouse anti-Human, PE, Clone: MAB11, BD

Mouse Monoclonal Antibody

Marca:  BD Biosciences 559321

Detalles adicionales : Peso : 0.02000kg

 Ver más versiones de este producto

Código de producto. 15889698

  • 576.00€ / cada uno
Fecha estimada de envío: 20-09-2024
para ver el stock

Descripción

Descripción

The MAb11 antibody reacts with human tumor necrosis factor (TNF, formerly known as TNF-α) protein. TNF is an efficient paracrine and endocrine mediator of inflammatory and immune functions. It regulates the growth and differentiation of a variety of cell types. TNF is cytotoxic for transformed cells when in conjunction with IFN-γ. It is secreted by activated monocytes/macrophages and other cells such as B cells, T cells and fibroblasts. The immunogen used to generate the MAb11 hybridoma was recombinant human TNF. The use of the MAb11 antibody has been reported to cross-react with TNF of rhesus monkey. This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Intracellular Staining

Especificaciones

Especificaciones

TNF
Monoclonal
PE
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Flow Cytometry
MAB11
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Recombinant Human TNF
100 Tests
Immunology
Human
IgG1 κ
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.