Learn More
TSG101 [Tumor Susceptibility Gene 101] Mouse, Unlabeled, Clone: 51, BD
Mouse Monoclonal Antibody
350.00€ - 565.00€
Especificaciones
Antígeno | TSG101 [Tumor Susceptibility Gene 101] |
---|---|
Clon | 51 |
Concentración | 250μg/mL |
Aplicaciones | Western Blot |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15886259
|
BD Biosciences
612696 |
50 μg |
350.00€
cada uno |
Fecha estimada de envío: 21-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15896259
|
BD Biosciences
612697 |
150 μg |
565.00€
cada uno |
Fecha estimada de envío: 12-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
Tumor Susceptibility Gene 101 (TSG101) was identified in a random mutagenesis screen for potential tumor suppressors in NIH 3T3 cells. Altered transcripts of this gene have been detected in sporadic breast cancers and many other human malignancies. Though the function of TSG101 is not clearly understood, its protein structure includes motifs involved in transcription regulation, and TSG101 has been shown to modulate the activation of steroid hormone receptors. In addition, TSG101 may have a role in regulating ubiquination. The N-terminal region of TSG101 contains a domain that resembles the catalytically active region of ubiquitin conjugases. However, TSG101 lacks an active-site cysteine crucial to the function of these conjugases. Interestingly, TSG101 interferes with MDM2 ubiquination leading to a decrease in MDM2 decay and down-regulation of p53 protein. Thus, TSG101 may function as a dominant-negative inhibitor of ubiquination in pathways where protein expression is tightly regulated.
Western Blotting
Especificaciones
TSG101 [Tumor Susceptibility Gene 101] | |
250μg/mL | |
Monoclonal | |
Mouse | |
Cell Biology | |
Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
IgG1 | |
Affinity Purified | |
Tumor Susceptibility Gene 101 |
51 | |
Western Blot | |
Unconjugated | |
RUO | |
Canine, Human, Mouse, Rat | |
Human TSG101 aa. 229-319 | |
Primary | |
Store undiluted at -20°C. |
For Research Use Only.