missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSPAN8 Monoclonal Antibody (458811), Invitrogen™
Rat Monoclonal Antibody
Marca: Invitrogen MA524179
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Reconstitute at 0.5 mg/mL in sterile PBS.
TSPAN8 Monoclonal antibody specifically detects TSPAN8 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot.Especificaciones
TSPAN8 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P19075 | |
Tspan8 | |
NS0 mouse myeloma cell line transfected with human TSPAN8 Met1-Lys237 (Ile35Val) | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
Flow Cytometry, Immunocytochemistry, Western Blot | |
458811 | |
Unconjugated | |
Tspan8 | |
C76990; CO-029; CO-29; D6.1A; E330007O21Rik; tetraspanin 8; Tetraspanin8; tetraspanin-8; Tm4sf3; transmembrane 4 superfamily member 3; TSPAN8; Tspan-8; Tumor-associated antigen CO-029 | |
Rat | |
Protein A/G | |
RUO | |
7103 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |