missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZO-1 Mouse anti-Human, Unlabeled, Clone: 1, BD
Mouse Monoclonal Antibody
357.00€ - 360.65€
Especificaciones
| Antígeno | ZO-1 |
|---|---|
| Clon | 1 |
| Concentración | 250μg/mL |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|---|---|---|---|---|---|---|---|---|---|
| Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
|
15895729
|
BD Biosciences
610966 |
50 μg |
357.00€
1 set |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
|
15805739
|
BD Biosciences
610967 |
150 μg |
361.00€ 360.65€ / 1 set Ahorro 0.35€ 0.10% Descuento |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | |||||
Descripción
Tight junctions ( z onulae o ccludens) are critical to the maintenance of cell polarity and intracellular barriers between epithelial and endothelial cells. Protein components of the tight junctions include actin filaments, symplekin, occludin, Rab3B, AF-6, 7H6, ZO-1, and ZO-2. Analysis of ZO-1 and -2 resulted in their inclusion in the MAGUK protein family. This family also includes the discs large tumor suppressor protein (Dlg-A) of Drosophila ; p55, an erythrocyte membrane protein; and PSD-95/SAP90, a synaptic membrane protein. All family members contain a region homologous to guanylate kinase (GuK), a src homology (SH3) domain, and multiple PDZ domains. Through these elements, MAGUK proteins function in signal transduction and, possibly, tumor suppression. ZO-1 is a peripheral membrane phosphoprotein that binds to other tight junction proteins such as occludin and AF-6. Via its SH3 domain, ZO-1 interacts with a serine protein kinase that phosphorylates a region immediately C-terminal of the SH3 domain. Taken together, these data indicate that ZO-1 is a critical element in the formation of tight junctions and may also serve an important role in signaling and tumor suppression.Especificaciones
| ZO-1 | |
| 250μg/mL | |
| Monoclonal | |
| Mouse | |
| Cell Biology | |
| Aqueous buffered solution containing BSA, glycerol, and ≤0.09% sodium azide. | |
| IgG1 | |
| Affinity Purified |
| 1 | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| Human ZO-1 aa. 1048-1247 | |
| Primary | |
| Store undiluted at -20°C. |
For Research Use Only.
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto