missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ AP2S1 (Human) Recombinant Protein
Human AP2S1 full-length ORF ( AAH06337, 1 a.a. - 142 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00001175-P01.25ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
- Sequence: MIRFILIQNRAGKTRLAKWYMQFDDDEKQKLIEEVHAVVTVRDAKHTNFVEFRNFKIIYRRYAGLYFCICVDVNDNNLAYLEAIHNFVEVLNEYFHNVCELDLVFNFYKVYTVVDEMFLAGEIRETSQTKVLKQLLMLQSLE
Especificaciones
AAH06337 | |
adaptor-related protein complex 2, sigma 1 subunit | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AP2S1 | |
GST |
1175 | |
Wheat germ expression system | |
25 μg | |
AP17, AP17-DELTA, CLAPS2 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |