missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APAF1 Monoclonal Antibody (5E11), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA515744
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
A suggested positive control is K562 cell lysate.
APAF1 gene encodes a cytoplasmic protein that initiates apoptosis. This protein contains several copies of the WD-40 domain, a caspase recruitment domain (CARD), and an ATPase domain (NB-ARC). Upon binding cytochrome c and dATP, this protein forms an oligomeric apoptosome. The apoptosome binds and cleaves caspase 9 preproprotein, releasing its mature, activated form. Activated caspase 9 stimulates the subsequent caspase cascade that commits the cell to apoptosis. Alternative splicing results in several transcript variants encoding different isoforms.Especificaciones
APAF1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.02% sodium azide | |
O14727, O88879, Q9EPV5 | |
APAF1 | |
A peptide corresponding to amino acids near the carboxy terminus of human Apaf1. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG1 κ |
Immunohistochemistry | |
5E11 | |
Unconjugated | |
APAF1 | |
6230400I06Rik; APAF1; APAF-1; Apaf1l; apoptotic peptidase activating factor 1; apoptotic protease activating factor 1; apoptotic protease-activating factor 1; CED4; DKFZp781B1145; fog; forebrain overgrowth; KIAA0413; mKIAA0413 | |
Mouse | |
Antigen affinity chromatography | |
RUO | |
11783, 317, 78963 | |
-20°C | |
Liquid |