Learn More
ApoA1 Monoclonal Antibody (513), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA514670
Descripción
By sandwich ELISA, MA5-14670 can be used as a detection antibody with Product # 710263 or # 701239 as a coating antibody, to generate a matched pair. Using these matched pairs, recombinant human Apo A-1, but not recombinant mouse Apo A-1, was detected. MA5-14670 can be used to detect Apo A-1 from serum samples. To increase sensitivity of sandwich ELISAs with MA5-14670, a biotinylated detection antibody followed by Streptavidin-HRP is recommended. By Western blot, MA5-14670 detects recombinant human Apo A-1, but not recombinant mouse Apo A-1. MA5-14670 is also recommended for detecting endogenous Apo A-1 by Western blot. Product MA514670 is a smaller package size of MIA1404 (formerly sold as a Seradyn product).
Apolipoprotein A-1 (apo A-1) in human plasma is a major protein component of high density lipoprotein (HDL). The molecular mass is 24 kDa. Apo A-1 is a cofactor for enzymes involved in the normal turnover of cholesterol and is produced in the liver. Plasma concentration is 270-380 mg/100 mL.![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Especificaciones
ApoA1 | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4 | |
P02647 | |
APOA1 | |
Purified human plasma Apo A-1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Radioimmune Assays (RIA), Western Blot | |
513 | |
Unconjugated | |
APOA1 | |
1-Sep; 2-Sep; Alp-1; Apo a; APO A1; apo A-1; APO I; apo(a); APOA1; Apoa-1; ApoAI; Apo-AI; apoA-I; Apolipoprotein; Apolipoprotein A1; apolipoprotein A-1; Apolipoprotein AI; apolipoprotein A-I; apolipoprotein A-I precursor; Apolipoprotein A-I(1-242); Brp-14; Ltw-1; Lvtw-1; MGC102525; MGC117399; MGC124929; preproapolipoprotein A-I; prepro-apolipoprotein-AI; ProapoA-I; Proapolipoprotein A-I; Sep-1; Sep2; Sep-2; Truncated apolipoprotein A-I; Udpgt; Ugt1; Ugt1a7 | |
Mouse | |
Ion-exchange chromatography | |
RUO | |
335 | |
Maintain refrigerated at 2-8°C for up to 6 months. For long term storage store at -20°C | |
Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.