missing translation for 'onlineSavingsMsg'
Learn More
Learn More
APOL1 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 11486-2-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody recognizes the endogenous APOL1 of 39-45 kDa in blood lysate.
Apolipoproteins are protein components of plasma lipoproteins. The apolipoprotein L gene family encodes six highly homologous proteins designated apoL-I to -VI, which are associated with large high density type lipoproteins (HDL). The human apoL family maps to chromosome 22q12.1- 13.1 within a 127,000-bp region. apoL has been characterized as a pancreas specific, 383-amino acid, 42 kDa protein that contains a 12-amino acid secretory signal peptide. The apoL genes have TATA-less promoters and contain putative sterol regulatory elements, suggesting that transcription of these genes may be coordinated with that of the low density lipoprotein receptor and genes in pathways involving the synthesis of triglycerides and cholesterol. apoL homologs can undergo 10 fold changes in expression during atherosclerotic changes in vascular endothelial cells, which includes the inflammatory reaction of atherosclerotic lesions.Especificaciones
APOL1 | |
Polyclonal | |
Unconjugated | |
APOL1 | |
APO L, APOL, APOL I, APOL1, Apolipoprotein L, Apolipoprotein L I, apolipoprotein L, 1, Apolipoprotein L1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
8542 | |
-20°C | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation | |
0.44 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
O14791 | |
APOL1 | |
APOL1 Fusion Protein Ag2016 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |