missing translation for 'onlineSavingsMsg'
Learn More

Apolipoprotein B/ApoB Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 6 publications

Marca:  Novus Biologicals NB200-527

Código de producto. 18721674

  • ONLINE EXCLUSIVE PRICE
    360.00€
    340.20€ / 0.50 ml
    Ahorro 19.80€
    5% Descuento

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

Apolipoprotein B/ApoB Polyclonal specifically detects Apolipoprotein B/ApoB in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
TRUSTED_SUSTAINABILITY
Especificaciones

Especificaciones

Apolipoprotein B/ApoB
Polyclonal
Western Blot 1:500, Immunoprecipitation
Apo B-100, apoB-100, apoB-48, apolipoprotein B (including Ag(x) antigen), apolipoprotein B-100, apolipoprotein B48, FLDB, LDLCQ4, mutant Apo B 100
Rabbit
Unpurified
RUO
Primary
Recognizes mouse as well as human Apo B48 and B100 as these proteins are highly conserved.
Store at -70C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunoprecipitation
Unconjugated
Whole antisera with 0.02% Sodium Azide
APOB
Mouse low density lipoprotein (LDL)
0.5 mL
Lipid and Metabolism
338
Human, Mouse
Antisera
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only