missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Artemin Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA5122226
135.34 EUR hasta el 2025-03-29
Use el código promocional "24111" para aplicar la oferta.
Alertas:
Para beneficiarse del descuento, los clientes deben comprar tres unidades del mismo producto al precio de catálogo en un solo pedido para recibir el 33.33% de descuento No hay límite para los múltiplos de 3 que los clientes pueden comprar PCODE:24111
Descripción
The protein encoded by this gene is a member of the glial cell line-derived neurotophic factor (GDNF) family of ligands which are a group of ligands within the TGF-beta superfamily of signaling molecules. GDNFs are unique in having neurotrophic properties and have potential use for gene therapy in neurodegenerative disease. Artemin has been shown in culture to support the survival of a number of peripheral neuron populations and at least one population of dopaminergic CNS neurons. Its role in the PNS and CNS is further substantiated by its expression pattern in the proximity of these neurons. This protein is a ligand for the RET receptor and uses GFR-alpha 3 as a coreceptor. Multiple transcript variants encoding different isoforms have been found for this gene.
Especificaciones
Artemin | |
Polyclonal | |
Unconjugated | |
ARTN | |
Artemin; ARTN; Enovin; EVN; NBN; neublastin; neurotrophic factor | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11876 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
500 μg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9Z0L2 | |
ARTN | |
Recombinant protein Artemin. The antigen corresponds to amino acid range 112-224 of the target protein. | |
20 μL | |
Primary | |
Mouse | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Artemin Polyclonal Antibody, Invitrogen™ > 20 μL; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido