missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Arylsulfatase B Monoclonal Antibody (708401), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA524327
Detalles adicionales : Peso : 0.00010kg
Descripción
Reconstitute in sterile PBS to a final concentration of 0.5 mg/mL.
Arylsulfatase B Monoclonal antibody specifically detects Arylsulfatase B in Human samples. It is validated for Immunocytochemistry, Western Blot.Especificaciones
Arylsulfatase B | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
P15848 | |
ARSB | |
CHO-derived recombinant human Arylsulfatase B/ARSB Ser37-Met533 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunocytochemistry, Western Blot | |
708401 | |
Unconjugated | |
ARSB | |
1110007C02Rik; AI480648; Arsb; Arylsulfatase B; Arylsulfatase B (MPS VI); arylsulfatase B complex; arylsulfatase B regulation; arylsulfatase B structural; arylsulfatase B temporal regulation; As1; As-1; As-1r; As1-r; As-1s; As1-s; As-1t; As1-t; ASB; Asr-1; Ast-1; G4S; MPS6; N-acetylgalactosamine-4-sulfatase | |
Mouse | |
Protein A/G | |
RUO | |
411 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |