missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ASL Monoclonal Antibody (OTI3D4), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA526531
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
ASL encodes a member of the lyase 1 family. The encoded protein forms a cytosolic homotetramer and primarily catalyzes the reversible hydrolytic cleavage of argininosuccinate into arginine and fumarate, an essential step in the liver in detoxifying ammonia via the urea cycle. Mutations in this gene result in the autosomal recessive disorder argininosuccinic aciduria, or argininosuccinic acid lyase deficiency. A nontranscribed pseudogene is also located on the long arm of chromosome 22. Alternatively spliced transcript variants encoding different isoforms have been described.Especificaciones
ASL | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P04424 | |
ASL | |
Full length human recombinant protein of ASL produced in HEK293T cell | |
100μL | |
Primary | |
Human | |
Antibody | |
IgG2a |
Immunohistochemistry (Paraffin), Western Blot | |
OTI3D4 | |
Unconjugated | |
ASL | |
2510006M18Rik; argininosuccinase; argininosuccinate lyase; Arginosuccinase; ASAL; Asl | |
Mouse | |
Affinity Chromatography | |
RUO | |
435 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido