Learn More
Tocris Bioscience™ ASP 3026
Potent anaplastic lymphoma kinase (ALK) inhibitor; also potent ACK inhibitor
Marca: Tocris Bioscience™ 5310/50
Detalles adicionales : CAS : 1097917-15-1 Peso : 0.00970kg
Descripción
ASP 3026 is a potent anaplastic lymphoma kinase (ALK) inhibitor (IC50 = 3.5 nM). Also inhibits Ack and ROS1 activity (IC50 values are 5.8 and 8.9 nM respectively). Attenuates proliferation, and induces apoptosis of NPM-ALK+ T cell anaplastic large-cell lymphoma (ALCL) cells in vitro. Suppresses tumor growth of NPM-ALK+ ALCL cell xenografts in mice. Causes tumor shrinkage in hEML4-ALK transgenic mice. Orally available.Especificaciones
N2-[2-Methoxy-4-[4-(4-methyl-1-piperazinyl)-1-piperidinyl]phenyl]-N4-[2-[(1-methylethyl)sulfonyl]phenyl]-1,3,5-triazine-2,4-diamine | |
1097917-15-1 | |
50 mg |