Learn More
TubeWriter Automated Labeling System
High speed automated labeling system
Marca: TubeWriter TW400A
Detalles adicionales : Peso : 36.00000kg
Descripción
- Full feature instrument for printing directly on curved surface, including tubes, vials, slides, plates and custom labware.
Especificaciones
Labeling System | |
TubeWriter 360™ | |
Not Autoclavable |