Learn More
Tocris Bioscience™ AZD 8055
Highly potent and selective mTOR inhibitor
Marca: Tocris Bioscience™ 7840/10
Detalles adicionales : CAS : 1009298-09-2 Peso : 0.00970kg
Descripción
AZD 8055 is a highly potent, selective and ATP-competitive mTOR inhibitor (IC50 = 0.8 nM). Inhibits growth and induces autophagy in a range of cancer cell lines and xenografts, including NSCLC and glioblastoma (IC50 values are 20, 50 and 53 nM in H838, A549 and U87MG cell lines, respectively). Orally bioavailable.Especificaciones
5-[2,4-Bis[(3S)-3-methyl-4-morpholinyl]pyrido[2,3-d]pyrimidin-7-yl]-2-methoxybenzenemethanol | |
10 mg | |
C25H31N5O4 |
1009298-09-2 | |
mTOR Inhibitors | |
0.98 |