missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BACE1 (beta-Secretase) Recombinant Protein
Click to view available options
Cantidad:
5 unidades
Tamaño de la unidad:
5 microgramos
Descripción
For maximum recovery please spin prior to use. Unless noted below, aliquots of the 5 μg, 10 μg and 20 μg sizes of kinase are not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots, aliquots below 20 μL are not recommended. Please never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase.
BACE1 (beta-secretase) is a key enzyme involved in the production of amyloid beta-peptides (Abeta) found in extracellular amyloid plaques of Alzheimer's disease (AD). In some cases early onset familial AD can be attributed to a Swedish mutation in the amyloid precursor protein (APP), which dramatically enhances the cleavage of this protein by BACE1. This and other genetic and pathological evidence has led to therapeutic approaches that have focused on the inhibition of BACE1 and other APP-cleaving enzymes, such as gamma-secretase stuff. APP is first cleaved by b-secretase, producing a soluble derivative of the protein and a membrane anchored 99-amino acid carboxy-terminal fragment (C99). The C99 fragment serves as substrate for g-secretase to generate the 4 kDa amyloid-b peptide, which is deposited in the brains of all sufferers of Alzheimer's disease. The long-sought b-secretase was recently identified by several groups independently and designated beta-site APP cleaving enzyme (BACE) and aspartyl protease 2 (Asp2). bACE/Asp2 is a novel transmembrane aspartic protease and colocalizes with APP.
Especificaciones
Especificaciones
| Número de acceso | P56817 |
| Concentración | See Label |
| Para utilizar con (aplicación) | Kinase Assay |
| Formulación | Proprietary buffer with no preservative |
| ID de gen (Entrez) | 23621 |
| Peso molecular | 55 kDa |
| Nombre | Human BACE1 (beta-Secretase) |
| Método de purificación | Purified |
| Cantidad | 5 unidades |
| Requisitos de almacenamiento | -80°C, Avoid Freeze/Thaw Cycles |
| Mostrar más |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido