missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bax Antibody (BAX/8960R) [Janelia Fluor™ 635], Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals (Bio-Techne) NBP3-23999JF635
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Bax Monoclonal antibody specifically detects Bax in Human samples. It is validated for Immunohistochemistry (Paraffin)Especificaciones
Bax | |
Monoclonal | |
Janelia Fluor 635 | |
apoptosis regulator BAX, BCL2-associated X protein, Bcl2-L-4, BCL2L4bcl2-L-4, Bcl-2-like protein 4 | |
Recombinant full-length human Bax protein | |
0.1 mL | |
Apoptosis, Cancer, Core ESC Like Genes, Stem Cell Markers, Tumor Suppressors | |
581 | |
Store at 4°C in the dark. | |
IgG κ |
Immunohistochemistry (Paraffin) | |
BAX/8960R | |
50mM Sodium Borate | |
Rabbit | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |